Learn More
Leinco Technologies Inc Anti-Human CD45RO (Clone UCHL-1) – Purified in vivo Functional GOLDTM Functional Grade
SDP
Supplier:  Leinco Technologies Inc C1105100MG
Encompass_Preferred
CD45 is a 180-240kD glycoprotein member of the protein tyrosine phosphatase (PTP) family known for its involvement in regulating a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. CD45 and its isoforms are vital regulators of T- and B-cell antigen receptor signaling. CD45 functions through its extracellular domain or through its cytoplasmic domain, and serves as a negative regulator of cytokine receptor signaling via JAK kinase supression. The large extracellular domain is highly glycosylated, and its multiple isoforms allow extensive variation in the structure of its side chains. CD45 isoforms show cell-type and differentiation-stage specific expression that can be used as markers that identify and distinguish between different types of immune cells. CD45R is an isoform of CD45 with a molecular weight of 220 kD.
Catalog No. 50-254-4260