Learn More
US Plastic Corporation 100 ML GRADUATED CYLINDERS
SDP
Catalog No. NC9244983
Encompass
Graduated cylinders have a Total Contained, Total Delivered single scale with raised numerals for easy reading. Flared top rims and large spouts facilitate pouring, and hexagon shaped bases provide stability.
Catalog No. NC9244983 Supplier US Plastic Corporation Supplier No. 70049
Graduated cylinders have a Total Contained, Total Delivered single scale with raised numerals for easy reading. Flared top rims and large spouts facilitate pouring, and hexagon shaped bases provide stability.