Learn More
Cpi International Sample Tubes, ModBlock, 100mL, Polypropylene, Hinged Caps, 250/pk
SDP
Catalog No. NC0703879
Encompass
100mL disposable digestions tubes for ModBlock. Tubes are graduated and have an attached hinged-cap
Catalog No. NC0703879 Supplier Cpi International Supplier No. 4370010202
100mL disposable digestions tubes for ModBlock. Tubes are graduated and have an attached hinged-cap