Learn More
Andwin Scientific 100 ML SOLUTION BASIN (80PK)
SDP
Catalog No. NC9513531
Encompass_Preferred
The pipettor basin is a disposable item that is compatible to use with 8 and 12 channel pipettors. With a capacity of 100 mL and a "V" shaped bottom, this product provides almost complete solution withdrawal. It is sterile, is free of RNase, DNase, and pyrogen, and is made of polystrene. This product is sold in a pack of 80.
Catalog No. NC9513531 Supplier Andwin Scientific Supplier No. 730014
The pipettor basin is a disposable item that is compatible to use with 8 and 12 channel pipettors. With a capacity of 100 mL and a "V" shaped bottom, this product provides almost complete solution withdrawal. It is sterile, is free of RNase, DNase, and pyrogen, and is made of polystrene. This product is sold in a pack of 80.