Learn More
Grainger Water Conditioner, 100ml Bottle
Catalog No. NC2001366
Encompass_Preferred
Manufactured by Bel Art Products. Distributed by Grainger. Protects water baths and other water systems against the growth of algae, bacteria, molds and biofilms. Protects pumps and thermostats and helps prevent tubing from clogging. Also permits longer use of water used in developer for color or X-ray films and will not harm film.
Catalog No. NC2001366 Supplier Grainger Supplier No. 8CJ34
Manufactured by Bel Art Products. Distributed by Grainger. Protects water baths and other water systems against the growth of algae, bacteria, molds and biofilms. Protects pumps and thermostats and helps prevent tubing from clogging. Also permits longer use of water used in developer for color or X-ray films and will not harm film.