Learn More
IBA LifeSciences 100 mM Biotin stock solution; 1 ml
Catalog No. 502100172
Encompass_Preferred
Biotin stock solution for preparing a dissociation buffer when using the Strep-tag technology for cells (Fab-TACS/Nano-TACS, CD3/CD28 Streptamer, MHC I Streptamer). Biotin addition causes the rapid dissociation of isolation reagents from the cell surface. Thus, fully functional, highly pure, and non-induced cells are received.
Catalog No. 50-210-0172 Supplier IBA LifeSciences Supplier No. 66325001
Biotin stock solution for preparing a dissociation buffer when using the Strep-tag technology for cells (Fab-TACS/Nano-TACS, CD3/CD28 Streptamer, MHC I Streptamer). Biotin addition causes the rapid dissociation of isolation reagents from the cell surface. Thus, fully functional, highly pure, and non-induced cells are received.