Learn More
Santa Cruz Biotechnology Glycine Buffer Solution, 100 mM
Catalog No. NC9085773
Encompass
For eluting antibodies in various antibody purification procedures, 0.2 µm filtered. pH: 2-2.5, Store at 4° C.
Catalog No. NC9085773 Supplier Santa Cruz Biotechnology Supplier No. SC295018
For eluting antibodies in various antibody purification procedures, 0.2 µm filtered. pH: 2-2.5, Store at 4° C.