Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metrohm USA Oxalic acid 100 MM | 144-62-7 | MFCD00002573 | 1 L
SDP

Catalog No. NC0498941 Shop All Metrohm USA Products
Encompass

Oxalic acid 100 MM | Mol Wt: 90.03 | 144-62-7 | MFCD00002573 | 1 L

Catalog No. NC0498941 Supplier Metrohm USA Supplier No. ERAIC1180
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxalic acid 100 MM | Mol Wt: 90.03 | 144-62-7 | MFCD00002573 | 1 L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.