Learn More
Andwin Scientific 100-PLACE HINGED FREEZER STORA
SDP
Supplier:  Andwin Scientific 23105800
Encompass_Preferred
With bold alpha-numeric markings, the 5 9/16" x 6" x 2 3/16" 100-place expanded polystyrene box offers security and organization for 1.5 and 2.0 ml tubes and most 2.0 ml cryogenic vials. The boxes have lids that has a durable, rolling hinge that withstands freezer storage and a snap-clasp that keeps boxes closed. This product comes in a pack of 5 boxes in natural color .
Catalog No. 50-819-918