Learn More
LabScientific 100-place premium microscope slide boxes. (12 pack) Durable ABS plastic box closes securely with a rust-resistant nickel-plated clasp and pin hinges. Securely stores slides in grooved, labeled slots.
Catalog No. NC1627863
Encompass
100-place premium microscope slide boxes. (12 pack) Durable ABS plastic box closes securely with a rust-resistant nickel-plated clasp and pin hinges. Securely stores slides in grooved, labeled slots. The molded feet enable secure stacking for easy transport and storage. Both the lining and the slots are numbered to correspond to the slide inventory sheet. The molded labeling area contains both "project" and "date" organization options. Dimensions: L x W x H: 208 x 175 x 34 mm
Catalog No. NC1627863 Supplier LabScientific Supplier No. SB100
100-place premium microscope slide boxes. (12 pack) Durable ABS plastic box closes securely with a rust-resistant nickel-plated clasp and pin hinges. Securely stores slides in grooved, labeled slots. The molded feet enable secure stacking for easy transport and storage. Both the lining and the slots are numbered to correspond to the slide inventory sheet. The molded labeling area contains both "project" and "date" organization options. Dimensions: L x W x H: 208 x 175 x 34 mm