Learn More
Electron Microscopy Sciences 100 Place Scintilation Vial Rack
SDP
Catalog No. 50949633
Encompass_Preferred
Welded polypropylene rack holds 100 standard 20 ml scintillation vials. The rack is autoclavable and resists both nitric and hydrochloric acids. The hole measures 11⁄16" diameter, dimensions: 14.5"(L) x 15"(W) x 2.5"(H)
Catalog No. 50-949-633 Supplier Electron Microscopy Sciences Supplier No. 72639
Welded polypropylene rack holds 100 standard 20 ml scintillation vials. The rack is autoclavable and resists both nitric and hydrochloric acids. The hole measures 11⁄16" diameter, dimensions: 14.5"(L) x 15"(W) x 2.5"(H)