Learn More
LabScientific 100-place premium tall microscope slide box (Blue). Additional space for those larger microscope slides (3x2"). Identification card, lining, and grooved numbered slots.
Catalog No. NC0054833
Encompass
100-place premium tall microscope slide box (Blue). Durable ABS plastic box closes securely with a rust-resistant nickel-plated clasp and pin hinges. Securely stores slides in grooved, labeled slots. The molded feet enable secure stacking for easy transport and storage. Both the lining and the slots are numbered to correspond to the slide inventory sheet. Dimensions: L x W x H: 208 x 175 x 60 mm
Catalog No. NC0054833 Supplier LabScientific Supplier No. SB10032B
100-place premium tall microscope slide box (Blue). Durable ABS plastic box closes securely with a rust-resistant nickel-plated clasp and pin hinges. Securely stores slides in grooved, labeled slots. The molded feet enable secure stacking for easy transport and storage. Both the lining and the slots are numbered to correspond to the slide inventory sheet. Dimensions: L x W x H: 208 x 175 x 60 mm