Learn More
Electron Microscopy Sciences Microscope Slide Box Microscope Slide Box 100 Place Tall
SDP
Catalog No. NC0815357
Encompass_Preferred
Specially designed for the 3 x 2 slides. This reinforced ABS material is rugged and designed not only for storage but transport of slides as well. Numbered grooved slots hold slides and cork liner ensures slides will not slip. Stackable. Stackable. The box measures 208 mm (L) x 175 mm (h) x 60 mm. The box has a capacity of 100 slides and is blue in color. FiloSlide and Supa Mega cassette compatible.
Catalog No. NC0815357 Supplier Electron Microscopy Sciences Supplier No. 41149
Specially designed for the 3 x 2 slides. This reinforced ABS material is rugged and designed not only for storage but transport of slides as well. Numbered grooved slots hold slides and cork liner ensures slides will not slip. Stackable. Stackable. The box measures 208 mm (L) x 175 mm (h) x 60 mm. The box has a capacity of 100 slides and is blue in color. FiloSlide and Supa Mega cassette compatible.