Learn More
Contec™ 100% Polyester Disposable String Mop Head
BIOPHARMA
Catalog No. 19084200 Shop All Contec Inc. Products
Change view
Click to view available options
Color:
Blue (Band)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-084-200 Blue (Band)
19-058-601 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-084-200 Supplier Contec™ Supplier No. C3MOP
Contec Disposable String Mop Head made from 100% polyester fabric, these mop heads absorb liquids quickly leaving mopped surfaces almost dry.
  • Provides excellent sorbency
  • Sterilizable by autoclave, ethylene oxide, or gamma radiation
  • Can be used with fiberglass handle
Specifications
Product Type Mop Head
Head Type String
Color Blue (Band)
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Single-use
Color-coded Yes
Material Non-woven Polyester