Learn More
Analytical Sales & Services 100 Position MicroCentrifuge Tube Storage Box, Blue
Catalog No. 5000804412
Encompass
Advantage™ 100 Position MicroCentrifuge Tube Storage Box with Polyurethane Foam Insert for 0.5mL - 2mL Tubes Color: Blue
Catalog No. 50-008-04412 Supplier Analytical Sales & Services Supplier No. 96315B
Advantage™ 100 Position MicroCentrifuge Tube Storage Box with Polyurethane Foam Insert for 0.5mL - 2mL Tubes Color: Blue