Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ 100% Recycled Polyethylene EcoPolyBlend™ Spill Control Pallet With Drain
GREENER_CHOICE

Catalog No. 19822471 Shop All Justrite Safety Group Products
Change view
Click to view available options
Material:
Recycled Polyethylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-822-471 Recycled Polyethylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-822-471 Supplier Justrite™ Supplier No. 28629
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Increase workplace safety and improve cleaniliness while using environmentally friendly products. The Justrite™ 100% Recycled Polyethylene EcoPolyBlend™ Spill Control Pallet With Drain helps to keep workers safe from accidental slips indoors and to avoid costly hazmat cleanup costs.

Made of recycled polyethylene, these pallets resist chemical attack from hazardous acids, caustics and solvents. The oversized sump holds the contents of a 55 gal. (200L) drum in the event of leak or rupture—meeting or exceeding the most stringent spill containment regulations. Seamless construction eliminates leaks. Self-locating grates lift off for quick clean up. Black pallets contain 100% recycled content—their dark color hides grime and dirt to keep your workplace looking clean. High visibility yellow pallets contain 35% recycled polyethylene.

Specifications

Color Black
Material Recycled Polyethylene
Dimensions (L x W x H) 25 x 11.6 x 73 in.
Width (English) 73 in.
Height (English) 11.6 in.
Depth (English) 25 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.