Learn More
Andwin Scientific 100-SLIDE BOXES 3X2 SLIDES100
SDP
Catalog No. 50931121
Encompass
The Large Glass Slide Storage Box can hold up to 100 slides measuring 3" x 2". The box has a sturdy wood frame that is covered with black embossed paper. Inside, there is a numbered plastic inert that gives spacing for the storage of the slides.
Catalog No. 50-931-121 Supplier Andwin Scientific Supplier No. 7148001
The Large Glass Slide Storage Box can hold up to 100 slides measuring 3" x 2". The box has a sturdy wood frame that is covered with black embossed paper. Inside, there is a numbered plastic inert that gives spacing for the storage of the slides.