Learn More
IMEB, Inc Tek-Select 100 Microscope Slide Storage Box, Plastic, White, High Impact Polystyrene & With Nickel Plated Rush-Resistant Clip, Each Lot Numbered & Corresponds to the Index on the Cover, Blue
SDP
Catalog No. 501689496
Encompass_Preferred
The Tek-Select 100 Microscope Slide Storage Box, Plastic, White, accommodates 100 slides 1 inx 3 in (25mm x 75mm) and are made of high impact polystyrene secured with nickel plated rush-resistant clip. Each lot is numbered and corresponds to the index on the cover to insure proper identification. The Tek-Select 100 slide box offers maximum protection for those slides requiring storage and transporting. Double hinged cover is attached to prevent the lid from becoming lost or broken.
Catalog No. 50-168-9496 Supplier IMEB, Inc Supplier No. HSBLB
The Tek-Select 100 Microscope Slide Storage Box, Plastic, White, accommodates 100 slides 1 inx 3 in (25mm x 75mm) and are made of high impact polystyrene secured with nickel plated rush-resistant clip. Each lot is numbered and corresponds to the index on the cover to insure proper identification. The Tek-Select 100 slide box offers maximum protection for those slides requiring storage and transporting. Double hinged cover is attached to prevent the lid from becoming lost or broken.