Learn More
Electron Microscopy Sciences 100 Slide Tray, Vertical
SDP
Supplier:  Electron Microscopy Sciences 7151001
Encompass_Preferred
Made from tough, resilient ASB plastic which offers long life. Holds 100-standard 1 x 3" slides positioned vertically for easy insertion in pre-numbered grooves allowing for simple identification and retrieval.
Catalog No. 50-931-153