Learn More
Bioassay Systems EnzyLight™ Cytotoxicity Assay Kit. For rapid, bioluminescent luciferin/luciferase based assay for cell viability, proliferation, cytotoxicity and high-throughput screen for anticancer agents.
SDP
Supplier:  Bioassay Systems ECTX100
Encompass_Preferred
For rapid, bioluminescent luciferin/luciferase based assay for cell viability, proliferation, cytotoxcity and high-throughput screen for anticancer agents. Key Features: Safe. Non-radioactive assay (cf. 3H-thymidine incorporation assay). Sensitive and accurate. As low as 50 cells can be quantified. Homogeneous and convenient. "Mix-incubate-measure" type assay. No wash and reagent transfer steps are involved. Robust and amenable to HTS: Z factors of 0.6 to 0.7 are routinely observed in 96-well and 384-well plates. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: Luminescence. Samples: Cells. Species: All. Procedure: Assay takes 2 min. Kit size: 100 tests. Detection limit: 50 cells.
Catalog No. 50-195-6301