Learn More
SAI Infusion Technologies Heparinized Saline Syringes with Plastic Needle - 0.5ml of 100 Units/ml Heparin in Saline, with attached plastic needle
SDP
Catalog No. NC0421592
Encompass
1ml Syringe filled with 0.5ml of 100 U/ml Heparin in 0.9% NaCl. Capped with a blunt plastic needle for accessing luer sites. 50 syringes per pack.
Maintain sterility in your preclinical studies with SAI's prefilled syringes. Our sterile prefilled syringes save time and money in preclinical studies requiring repeated catheter access. Product is for laboratory animal research only.
Catalog No. NC0421592 Supplier SAI Infusion Technologies Supplier No. HSC10010.5PN
1ml Syringe filled with 0.5ml of 100 U/ml Heparin in 0.9% NaCl. Capped with a blunt plastic needle for accessing luer sites. 50 syringes per pack.
Maintain sterility in your preclinical studies with SAI's prefilled syringes. Our sterile prefilled syringes save time and money in preclinical studies requiring repeated catheter access. Product is for laboratory animal research only.