Learn More
Trajan Scientific 100 UL EVOL SYRINGE
SDP
Catalog No. 502530067
Encompass_Preferred
Trajan eVol XCHANGE Syringes, Volume: 100 uL, Length Needle: 50 mm, Needle Point Style: Bevel, Needle Gauge: 25 G, For Use With: Liquid handling procedure, Length: 1.9 in., 50 mm, eVol Syringe
Catalog No. 50-253-0067 Supplier Trajan Scientific Supplier No. 2910029
Trajan eVol XCHANGE Syringes, Volume: 100 uL, Length Needle: 50 mm, Needle Point Style: Bevel, Needle Gauge: 25 G, For Use With: Liquid handling procedure, Length: 1.9 in., 50 mm, eVol Syringe