Learn More
Thermo Scientific Vanquish Split Sampler Sample Loops | Stainless Steel | 1 EA
Catalog No. NC2099249 Shop All Thermo Scientific Products
Encompass
Automate sample injection into your Thermo Scientific Vanquish HPLC or UHPLC system, Available in multiple sizes (10, 25, 100, 250, 1000 µL), Viper Fittings included for easy installation with leak-free connections, Biocompatible versions available
This item is not returnable. View return policy
Catalog No. NC2099249 Supplier Thermo Scientific Supplier No. 6851.1950
Automate sample injection into your Thermo Scientific Vanquish HPLC or UHPLC system, Available in multiple sizes (10, 25, 100, 250, 1000 µL), Viper Fittings included for easy installation with leak-free connections, Biocompatible versions available