Learn More
Electron Microscopy Sciences PCR 100-Well Storage Rack, Natural
SDP
Catalog No. 5034140
Encompass_Preferred
Polypropylene rack features durable three point hinges and clasp for secure lid closure. Holds 1.5 to 2.0 ml microtubes and cryovials. User friendly rack features molded gridlines on the lid, imprinted coordinates on the bottom of base, and imprinted and raised coordinates on the box. Rack measures 5.625 x 5.875 x 2.25".
Color: Natural
Catalog No. 50-341-40 Supplier Electron Microscopy Sciences Supplier No. 61601N
Polypropylene rack features durable three point hinges and clasp for secure lid closure. Holds 1.5 to 2.0 ml microtubes and cryovials. User friendly rack features molded gridlines on the lid, imprinted coordinates on the bottom of base, and imprinted and raised coordinates on the box. Rack measures 5.625 x 5.875 x 2.25".
Color: Natural