Learn More
Bellco Glass, Inc. 100 x 15 mm Petri Dish, X-Plate (4-section), stackable, sterile, 20/pk, 500/cs
SDP
Catalog No. 502330703
Encompass_Preferred
100 x 15 mm Petri Dish, X-Plate (4-section), stackable, sterile, 20/pk, 500/cs. These non-treated polystyrene plates are transparent and DNase/RNase-Free. The bottom has a ridge allowing for easy handling and stacking. The plate has three vents for gas exchange. • High clarity polystyrene• Flat, transparent surface• Stackable for easy storage and handling• Sterilized by E-Beam• Non-pyrogenic
Catalog No. 50-233-0703 Supplier Bellco Glass, Inc. Supplier No. 451400479
100 x 15 mm Petri Dish, X-Plate (4-section), stackable, sterile, 20/pk, 500/cs. These non-treated polystyrene plates are transparent and DNase/RNase-Free. The bottom has a ridge allowing for easy handling and stacking. The plate has three vents for gas exchange. • High clarity polystyrene• Flat, transparent surface• Stackable for easy storage and handling• Sterilized by E-Beam• Non-pyrogenic