Learn More
JDM LAB SOLUTIONS LLC Oxford Lab Products Pipette Tips, 1000 µL, Low Retention, LTS Compatible, 4800/Cs
Catalog No. NC3935290
Encompass
These 1000 µL pipette tips are precision-engineered for compatibility with LTS style pipettes, including both single and multi-channel variants. Manufactured from high-quality, FDA-approved virgin polypropylene using diamond-finished molds, these tips feature an ultra-fine point and a hydrophobic resin coating to minimize sample retention and ensure high-precision liquid delivery.
  • Compatible with LTS and XLS style pipettes
  • Low-retention internal surface reduces liquid hold-back
  • Certified free of RNase, DNase, PCR inhibitors, endotoxins, and pyrogens
  • Graduation marks for visual volume verification
  • Minimal attachment force required for secure sealing
  • Manufactured in a touch-free, automated facility
  • Available in eco-friendly reload stack packaging
Catalog No. NC3935290 Supplier JDM LAB SOLUTIONS LLC Supplier No. LTR1000LR
These 1000 µL pipette tips are precision-engineered for compatibility with LTS style pipettes, including both single and multi-channel variants. Manufactured from high-quality, FDA-approved virgin polypropylene using diamond-finished molds, these tips feature an ultra-fine point and a hydrophobic resin coating to minimize sample retention and ensure high-precision liquid delivery.
  • Compatible with LTS and XLS style pipettes
  • Low-retention internal surface reduces liquid hold-back
  • Certified free of RNase, DNase, PCR inhibitors, endotoxins, and pyrogens
  • Graduation marks for visual volume verification
  • Minimal attachment force required for secure sealing
  • Manufactured in a touch-free, automated facility
  • Available in eco-friendly reload stack packaging