Learn More
CELLTREAT Scientific Products 1000µL Pipette Tips, LfTS Fit, Non-sterile
SDP
Catalog No. 5000144326
Encompass_Preferred
Tips are designed to provide a more comfortable pipetting experiencePipette tips are packaged in racks with hinged lids designed for easy one-handed opening10μl tips feature an extended length for easier access to the bottom of vesselsFilter tips are fitted with a polymer aerosol barrier to reduce the risk of contaminationLids feature stacking supports, and are removable to save bench space
Catalog No. 50-001-44326 Supplier CELLTREAT Scientific Products Supplier No. 229076NS
Tips are designed to provide a more comfortable pipetting experiencePipette tips are packaged in racks with hinged lids designed for easy one-handed opening10μl tips feature an extended length for easier access to the bottom of vesselsFilter tips are fitted with a polymer aerosol barrier to reduce the risk of contaminationLids feature stacking supports, and are removable to save bench space