Learn More
Perkin Elmer US LLC 1000 µL Syringe Kit with 2000 µL Buffer Tubing
Catalog No. 502112568
Encompass_Preferred
These glass syringes come with precision stainless steel plungers. They are used for sampling and flushing and are available in a large variety of sizes.
Catalog No. 50-211-2568 Supplier Perkin Elmer US LLC Supplier No. N2936054
These glass syringes come with precision stainless steel plungers. They are used for sampling and flushing and are available in a large variety of sizes.