Learn More
GENTECH SOLUTIONS LLC 1000µl Universal Grad Tip, Bulk, Sterile
Catalog No. NC3728775
Encompass_Preferred
EliteChem Ensure accurate, consistent results with our high-quality Pipette Tips, designed for a universal fit on all major pipette brands. Made from durable, high-quality polypropylene, they help maximize sample recovery and are available in sterile, RNase/DNase-free options for sensitive lab work. Choose from a variety of volumes and packaging styles to suit your workflow needs.
Catalog No. NC3728775 Supplier GENTECH SOLUTIONS LLC Supplier No. 1800150011
EliteChem Ensure accurate, consistent results with our high-quality Pipette Tips, designed for a universal fit on all major pipette brands. Made from durable, high-quality polypropylene, they help maximize sample recovery and are available in sterile, RNase/DNase-free options for sensitive lab work. Choose from a variety of volumes and packaging styles to suit your workflow needs.