Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 1000 μL Universal Pipetter Tips: Bevelled, Maxymum Recovery

Catalog No. 14222695 Shop All Axygen Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Format
14-222-695 Non-sterile Rack
14-222-696 Sterile Rack
14-222-694 Non-sterile Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 14-222-695 Supplier Axygen™ Supplier No. T1000CLR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Compatible with most popular brands of pipettes

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Beveled
  • Maxymum Recovery technology for ultra-low retention and high yield
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 100 to 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Rack Color Clear
Autoclavable Autoclavable
Model T-1000-C-L-R
Certifications/Compliance EN ISO 13485 compliant, BSE/TSE compliant, Non-Pyrogenic, USP Class VI tested, Human DNA Free, DNase/RNase Free, Quality Control Tested
Clean Claims DNase, RNase, and Pyrogen free
Format Rack
Tip Style Low Retention, Universal
Packaging Hinged rack
Quantity 1000/Pk., 5000/Cs.
Product Type Pipette Tips
Includes Tips, Rack, Pack, Case
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line Maxymum Recovery
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.