Learn More
Boston Bioproducts Inc Transfer Buffer (10X, Electro Blotting) - 1L
Catalog No. NC9115191
Encompass
The 10X Transfer Buffer-Electro Blotting is the most commonly used buffer in the wet transfer system for the electrotransfer of proteins from SDS-PAGE gels to nitrocellulose or PVDF membranes. The addition of 10% methanol in thhis buffer prevents the gel from swelling during transfer and enhances efficiency of low abundancy proteins and protein binding to nitrocellulose membrane. This buffer is also used as a running buffer for native protein gel electrophoresis.
Catalog No. NC9115191 Supplier Boston Bioproducts Inc Supplier No. BP1901L
The 10X Transfer Buffer-Electro Blotting is the most commonly used buffer in the wet transfer system for the electrotransfer of proteins from SDS-PAGE gels to nitrocellulose or PVDF membranes. The addition of 10% methanol in thhis buffer prevents the gel from swelling during transfer and enhances efficiency of low abundancy proteins and protein binding to nitrocellulose membrane. This buffer is also used as a running buffer for native protein gel electrophoresis.