Learn More
Genesee Scientific Corporation 1000µl Reach Ergonomic Filter Pipet Tips Low Binding, Racked, Sterile, 8 Racks of 96 Tips/Unit
SDP
Catalog No. NC9783028
Encompass
Olympus Plastics™ Pipet Tip designed for standard pipettors. Patented ergonomic design engineered for effortless insertion and ejection. Reduced risk of Repetitive Stress Disorder (RSD). Naturally low-retentive resin that maximizes sample recovery; no secondary chemicals
Catalog No. NC9783028 Supplier Genesee Scientific Corporation Supplier No. 24430
Olympus Plastics™ Pipet Tip designed for standard pipettors. Patented ergonomic design engineered for effortless insertion and ejection. Reduced risk of Repetitive Stress Disorder (RSD). Naturally low-retentive resin that maximizes sample recovery; no secondary chemicals