Learn More
Savillex Corporation 1000 ML STANDARD JAR,EACH
SDP
Catalog No. NC1921652
Encompass
•Service temperature range of PFA is ‑328°F to 500°F (‑200°C to 260°C)•Wide opening for easy retrieval of samples•Four ribbed areas for easy handling•Translucent
Catalog No. NC1921652 Supplier Savillex Corporation Supplier No. 100100001
•Service temperature range of PFA is ‑328°F to 500°F (‑200°C to 260°C)•Wide opening for easy retrieval of samples•Four ribbed areas for easy handling•Translucent