Learn More
Scytek Laboratories Super Block, 1000 mL, Serum-Free Protein Block
SDP
Catalog No. NC9782835
Encompass_Preferred
Unique serum-free protein block for immunochemistry. Eliminates the need for matching species with the link antibody and is often, more effective at reducing non-specific background staining than normal serum. Variety of applications.
Catalog No. NC9782835 Supplier Scytek Laboratories Supplier No. AAA999
Unique serum-free protein block for immunochemistry. Eliminates the need for matching species with the link antibody and is often, more effective at reducing non-specific background staining than normal serum. Variety of applications.