Learn More
SPEX CERTIPREP LLC 3P Silica (1000g/mL in H2O), 250mL
Supplier:  SPEX CERTIPREP LLC 3P 1000SIO24F250
Encompass_Preferred
(HPS by Zeptometrix) 1000 g/mL Silica (SiO2) from (NH4)2SiF6 in H2O. This standard is traceable to NIST SRM 3150 and certified ISO 17034 and 17025.
Catalog No. NC1700191