Learn More
SPEX CERTIPREP LLC 3P Nitrate from Sodium Nitrate at 1,000 ug/mL in H2O, 100 mL
Catalog No. NC1075622
Encompass_Preferred
(HPS by Zeptometrix) Nitrate Anion IC Single-Component Standard from Sodium Nitrate at 1,000 ug/mL in H2O. Can be used as stock solution for nitrate as nitrate, EPA Method 9056. If you need to report Nitrogen using a nitrate source, please see IC-NO3-M, 1,000 g/mL Nitrogen in H2O instead.
Catalog No. NC1075622 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICNOM100
(HPS by Zeptometrix) Nitrate Anion IC Single-Component Standard from Sodium Nitrate at 1,000 ug/mL in H2O. Can be used as stock solution for nitrate as nitrate, EPA Method 9056. If you need to report Nitrogen using a nitrate source, please see IC-NO3-M, 1,000 g/mL Nitrogen in H2O instead.