Learn More
Aqua Solutions 1000 PPM TOC STD 500ML
SDP
Catalog No. NC9583767
Encompass_Preferred
Total Organic Carbon Standard 1000mg/L(ppm) Solution Prepared to EPA 415/Standard Methods, Health: 1, Flammability: 0, Reactivity: 0, Specific Gravity: 1, Percent Volatile by Volume: >99, Solubility in Water: Soluble, Stability: Stable
Catalog No. NC9583767 Supplier Aqua Solutions Supplier No. 9570500ML
Total Organic Carbon Standard 1000mg/L(ppm) Solution Prepared to EPA 415/Standard Methods, Health: 1, Flammability: 0, Reactivity: 0, Specific Gravity: 1, Percent Volatile by Volume: >99, Solubility in Water: Soluble, Stability: Stable