Learn More
Analytical Sales & Services 1000 psi BPR Assembly with PEEK Holder and Fittings
Catalog No. 5000804874
Encompass
1000 psi BPR Assembly with PEEK Holder and Fittings\n\nQty/Pkg: EACH
Catalog No. 50-008-04874 Supplier Analytical Sales & Services Supplier No. 59455
1000 psi BPR Assembly with PEEK Holder and Fittings\n\nQty/Pkg: EACH