Learn More
IDEX Health & Science Upchurch 1000 PSI BPR CARTRIDGE
Catalog No. NC1919878
Encompass_Preferred
Idex Back Pressure Regulators: Cartridges, Pressure: 1000 psi, Swept Volume: 83 uL, For Use With: Back Pressure Regulators
Catalog No. NC1919878 Supplier IDEX Health & Science Upchurch Supplier No. P796
Idex Back Pressure Regulators: Cartridges, Pressure: 1000 psi, Swept Volume: 83 uL, For Use With: Back Pressure Regulators