Learn More
Chemglass Life Sciences Chemglass 1000 μL Automation Clear Tips, Sterile, Barcoded - 96 tips/rack, 10 racks/pack
SDP
Catalog No. 502733256
Encompass_Preferred
Chemglass 1000 μL automation clear pipette tips are sterile, DNase/RNase-free, medical-grade polypropylene tips designed for high-throughput liquid handling on automated workstations. Supplied in barcoded 96-tip racks, they provide reliable performance for genomics, proteomics, immunoassays, and other automated laboratory workflows.
  • 1000 μL tip volume.
  • Sterile; electron-beam sterilized.
  • DNase/RNase-free.
  • Medical-grade polypropylene construction.
  • Barcoded 96-tip racks for automation traceability.
  • 10 racks per pack; 50 packs per case.
  • Produced in a controlled clean-room environment.
  • Compatible with common automated liquid-handling workstations; verify fit with instrument.
Catalog No. 50-273-3256 Supplier Chemglass Life Sciences Supplier No. CLS3499A065
Chemglass 1000 μL automation clear pipette tips are sterile, DNase/RNase-free, medical-grade polypropylene tips designed for high-throughput liquid handling on automated workstations. Supplied in barcoded 96-tip racks, they provide reliable performance for genomics, proteomics, immunoassays, and other automated laboratory workflows.
  • 1000 μL tip volume.
  • Sterile; electron-beam sterilized.
  • DNase/RNase-free.
  • Medical-grade polypropylene construction.
  • Barcoded 96-tip racks for automation traceability.
  • 10 racks per pack; 50 packs per case.
  • Produced in a controlled clean-room environment.
  • Compatible with common automated liquid-handling workstations; verify fit with instrument.