Learn More
Chemglass Life Sciences Chemglass 1000 μL Automation clear tips for Hamilton, sterile, 96/pack (2,304/case)
SDP
Catalog No. 502733266
Encompass_Preferred
Sterile 1000 μL automation clear tips designed for Hamilton automated liquid-handling systems. Manufactured from medical-grade polypropylene and DNase/RNase-free, these nest-packaged tips are electron-beam sterilized and supplied with barcode identification for high-throughput workflows.
  • Volume: 1000 μL
  • Compatibility: Hamilton automated pipetting systems
  • Material: medical-grade polypropylene
  • Sterilization: electron beam sterilized
  • RNase/DNase status: DNase/RNase free
  • Packaging: nest blister packs, sterile
  • Count: 96 tips per pack, 2,304 tips per case
  • Barcode: included for inventory and automation
Catalog No. 50-273-3266 Supplier Chemglass Life Sciences Supplier No. CLS3499A045
Sterile 1000 μL automation clear tips designed for Hamilton automated liquid-handling systems. Manufactured from medical-grade polypropylene and DNase/RNase-free, these nest-packaged tips are electron-beam sterilized and supplied with barcode identification for high-throughput workflows.
  • Volume: 1000 μL
  • Compatibility: Hamilton automated pipetting systems
  • Material: medical-grade polypropylene
  • Sterilization: electron beam sterilized
  • RNase/DNase status: DNase/RNase free
  • Packaging: nest blister packs, sterile
  • Count: 96 tips per pack, 2,304 tips per case
  • Barcode: included for inventory and automation