Learn More
Chemglass Life Sciences Automation Tips for Tecan, 1,000 μL Clear Pipette Tips, Sterile, 96/rack, Boxed (10 racks/pack, 50 racks/case)
SDP
Catalog No. 502733233
Encompass_Preferred
Clear, 1,000 μL automation pipette tips designed for high-throughput liquid handling applications. Supplied sterile and rack-packaged for automated workflows, they offer contamination control and consistent volume transfer for reliable performance.
  • 1,000 μL capacity
  • Medical-grade polypropylene construction for chemical resistance
  • Sterile, DNase/RNase-free; electron-beam sterilized
  • Packaged 96 tips per rack, 10 racks per pack, 50 racks per case
  • Produced in a 100,008 grade clean room
  • Designed for automated liquid handling systems
Catalog No. 50-273-3233 Supplier Chemglass Life Sciences Supplier No. CLS3499A405
Add to Cart
Add to Cart
Clear, 1,000 μL automation pipette tips designed for high-throughput liquid handling applications. Supplied sterile and rack-packaged for automated workflows, they offer contamination control and consistent volume transfer for reliable performance.
  • 1,000 μL capacity
  • Medical-grade polypropylene construction for chemical resistance
  • Sterile, DNase/RNase-free; electron-beam sterilized
  • Packaged 96 tips per rack, 10 racks per pack, 50 racks per case
  • Produced in a 100,008 grade clean room
  • Designed for automated liquid handling systems