Learn More
Chemglass Life Sciences Chemglass 1,000 μL Automation Conductive Filter Tips for Hamilton Systems, Sterile, Boxed, 96/rack (CLS-3499A-033)
SDP
Catalog No. 502733270
Encompass_Preferred
These 1,000 μL automation conductive filter tips provide sterile, low-contamination liquid handling for automated platforms. Manufactured from medical-grade polypropylene and electron-beam sterilized in a cleanroom environment, the tips include aerosol-resistant filters and are rack-packed with barcodes for automation and inventory tracking.
  • 1,000 μL nominal volume.
  • Medical-grade polypropylene construction.
  • Sterile; electron beam sterilized.
  • DNase/RNase free.
  • Aerosol-resistant filter to reduce sample carryover.
  • Packaging: 96 tips per rack; 10 racks per pack; 50 racks per case.
  • Barcoded racks and boxes for automation tracking.
  • Boxed, nest format for robotic loading.
Catalog No. 50-273-3270 Supplier Chemglass Life Sciences Supplier No. CLS3499A033
These 1,000 μL automation conductive filter tips provide sterile, low-contamination liquid handling for automated platforms. Manufactured from medical-grade polypropylene and electron-beam sterilized in a cleanroom environment, the tips include aerosol-resistant filters and are rack-packed with barcodes for automation and inventory tracking.
  • 1,000 μL nominal volume.
  • Medical-grade polypropylene construction.
  • Sterile; electron beam sterilized.
  • DNase/RNase free.
  • Aerosol-resistant filter to reduce sample carryover.
  • Packaging: 96 tips per rack; 10 racks per pack; 50 racks per case.
  • Barcoded racks and boxes for automation tracking.
  • Boxed, nest format for robotic loading.