Learn More
Chemglass Life Sciences Automation Conductive Filter Tips for Hamilton, 1000 μL, Sterile, 96 Tips x 5 Blister Pack (3,840/case)
SDP
Catalog No. 502733274
Encompass_Preferred
Conductive 1,000 μL filter tips designed for high-throughput automated pipetting on Hamilton robotic systems. Electron beam sterilized and DNase/RNase free, the tips feature aerosol-resistant filters, barcode-labeled racks, and preloaded nested blister packaging to support reliable automation workflows.
  • High-volume 1,000 μL capacity.
  • Conductive design for robotic systems.
  • Aerosol-resistant filter to reduce contamination risk.
  • Preloaded in nested blister packs for easy loading.
  • Barcode-labeled racks for automation tracking.
  • Electron beam sterilized and DNase/RNase free.
  • Medical-grade polypropylene construction.
  • Pack configuration: 96 tips x 5 blister packs; 3,840 tips per case.
Catalog No. 50-273-3274 Supplier Chemglass Life Sciences Supplier No. CLS3499A025
Add to Cart
Add to Cart
Conductive 1,000 μL filter tips designed for high-throughput automated pipetting on Hamilton robotic systems. Electron beam sterilized and DNase/RNase free, the tips feature aerosol-resistant filters, barcode-labeled racks, and preloaded nested blister packaging to support reliable automation workflows.
  • High-volume 1,000 μL capacity.
  • Conductive design for robotic systems.
  • Aerosol-resistant filter to reduce contamination risk.
  • Preloaded in nested blister packs for easy loading.
  • Barcode-labeled racks for automation tracking.
  • Electron beam sterilized and DNase/RNase free.
  • Medical-grade polypropylene construction.
  • Pack configuration: 96 tips x 5 blister packs; 3,840 tips per case.