Learn More
Chemglass Life Sciences 1000 μL Robotic conductive filter tips for Tecan - sterile, 96/rack; 10 racks/pack; 5 packs/case (Nest 332216 / Chemglass CLS-3499A-093)
SDP
Catalog No. 502733252
Encompass_Preferred
Sterile 1000 μL conductive robotic filter tips engineered for high-throughput automated pipetting on Tecan liquid handlers. Medical-grade conductive polypropylene and aerosol-resistant filters minimize contamination risk and ensure consistent liquid handling performance in genomics, proteomics, and biopharmaceutical workflows.
  • Volume: 1000 μL.
  • Pack configuration: 96 tips per rack; 10 racks per pack; 5 packs per case (4,800 tips per case).
  • Material: conductive polypropylene.
  • Sterile: yes; e-beam sterilized to SAL 10^-6.
  • Filter type: aerosol-resistant filter to reduce contamination.
  • Compatibility: designed for common robotic liquid handlers and 96-position systems.
  • Manufacturing standard: produced under ISO 13485 processes.
Catalog No. 50-273-3252 Supplier Chemglass Life Sciences Supplier No. CLS3499A093
Add to Cart
Add to Cart
Sterile 1000 μL conductive robotic filter tips engineered for high-throughput automated pipetting on Tecan liquid handlers. Medical-grade conductive polypropylene and aerosol-resistant filters minimize contamination risk and ensure consistent liquid handling performance in genomics, proteomics, and biopharmaceutical workflows.
  • Volume: 1000 μL.
  • Pack configuration: 96 tips per rack; 10 racks per pack; 5 packs per case (4,800 tips per case).
  • Material: conductive polypropylene.
  • Sterile: yes; e-beam sterilized to SAL 10^-6.
  • Filter type: aerosol-resistant filter to reduce contamination.
  • Compatibility: designed for common robotic liquid handlers and 96-position systems.
  • Manufacturing standard: produced under ISO 13485 processes.