Learn More
USA Scientific Inc 1000 uL XL, LTS compatible
Supplier:  USA Scientific Inc 10703701
Encompass
1000 uL XL, LTS compatible tips for Rainin pipettes, non-filter, refill, non-str, low retention, 768 tips in 8 wafers
Catalog No. 50-274-5163