Learn More
Sino Biological Recombinant Human EGFR Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10001H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ser 645) of human EGFR (NP_005219) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-3718