Learn More
Sino Biological Recombinant Human EGFR Protein (ECD, His Tag), HPLC-verified
Catalog No. 501613723
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ser 645) of human EGFR (NP_005219) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-3723 Supplier Sino Biological Supplier No. 10001H08H100
A DNA sequence encoding the extracellular domain (Met 1-Ser 645) of human EGFR (NP_005219) was expressed with a C-terminal polyhistidine tag.