Learn More
Sino Biological Recombinant Human EGFR Protein (aa 668-1210, His & GST Tag)
Supplier:  Sino Biological 10001H20B250
Encompass_Preferred
A DNA sequence encoding the cytoplasmic domain (Met 668-Ala 1210) of human EGFR (NP_005219) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-3727