Learn More
Sino Biological Anti-EGFR Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10001MM08F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human EGFR / HER1 / ErbB1 (rh EGFR / HER1 / ErbB1; Catalog#10001-H08H; NP_005219; Met 1-Ser 645) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-161-0255